Merck
Amphétamines
Amphétamines suitable for GC, application for SPME
Matériaux
Fibre SPME
Réf. du produit
Description
Tarif
Fibres SPME Polydimethylsiloxane (PDMS)
df 100 μm(PDMS, needle size 24 ga, for use with manual holder
CONDITIONS
sample/matrix
1 mL urine + 0.7g potassium carbonate in 20 mL headspace vial, equilibrated at 80°C, 30 min
SPME fiber
polydimethylsiloxane, 100 μm (57300-U)
derivatization
methyl bis-trifluoroacetamide (headspace, 0.5 min, ambient)
extraction
headspace, 3-5 min, 80 °C
desorption process
1 min, 270 °C
column
100% methyl polysiloxane, 12.5 m × 0.2 mm I.D., 0.33 μm (Supelco equivalent, Equity-1, 28041-U)
oven
60 °C (1 min) to 140 °C (4 min) at 30 °C/min, then to 276 °C at 20 °C/min, 4 min
inj. temp.
splitless (closed 1 min), 270 °C
detector
MS, full scan