Merck
  • Mitotic arrest-induced phosphorylation of Mcl-1 revisited using two-dimensional gel electrophoresis and phosphoproteomics: nine phosphorylation sites identified.
Mitotic arrest-induced phosphorylation of Mcl-1 revisited using two-dimensional gel electrophoresis and phosphoproteomics: nine phosphorylation sites identified.
Oncotarget (2016-10-16)
Rong Chu, Sarah E Alford, Katherine Hart, Anisha Kothari, Samuel G Mackintosh, Matthew R Kovak, Timothy C Chambers
RÉSUMÉ
Microtubule targeting agents (MTAs) characteristically promote phosphorylation and degradation of Mcl-1, and this represents a critical pro-apoptotic signal in mitotic death. While several phosphorylation sites and kinases have been implicated in mitotic arrest-induced Mcl-1 phosphorylation, a comprehensive biochemical analysis has been lacking. Contrary to previous reports suggesting that T92 phosphorylation by Cdk1 regulates Mcl-1 degradation, a T92A Mcl-1 mutant expressed in HeLa cells was phosphorylated and degraded with the same kinetics as wild-type Mcl-1 following vinblastine treatment. Similarly, when Mcl-1 with alanine replacements of all five putative Cdk sites (S64, T92, S121, S159, T163) was expressed, it was also phosphorylated and degraded in response to vinblastine. To analyze Mcl-1 phosphorylation in more detail, two-dimensional gel electrophoresis (2D-PAGE) was performed. While untreated cells expressed mainly unphosphorylated Mcl-1 with two minor phosphorylated species, Mcl-1 from vinblastine treated cells migrated during 2D-PAGE as a train of acidic spots representing nine or more phosphorylated species. Immunopurification and mass spectrometry of phosphorylated Mcl-1 derived from mitotically arrested HeLa cells revealed nine distinct sites, including several previously unreported. Mcl-1 bearing substitutions of all nine sites had a longer half-life than wild-type Mcl-1 under basal conditions, but still underwent phosphorylation and degradation in response to vinblastine treatment, and, like wild-type Mcl-1, was unable to protect cells from MTA treatment. These results reveal an unexpected complexity in Mcl-1 phosphorylation in response to MTAs and indicate that previous work has severely underestimated the number of sites, and thus encourage major revisions to the current model.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3,4,4′-Trichlorocarbanilide, 99%