Merck
  • Effective adoptive immunotherapy by T-LAK cells retargeted with bacterial superantigen-conjugated antibody to MUC1 in xenografted severe combined immunodeficient mice.
Effective adoptive immunotherapy by T-LAK cells retargeted with bacterial superantigen-conjugated antibody to MUC1 in xenografted severe combined immunodeficient mice.
Cancer research (1998-07-14)
M Shinoda, T Kudo, M Suzuki, Y Katayose, N Sakurai, H Saeki, H Kodama, K Fukuhara, K Imai, Y Hinoda, S Matsuno
RÉSUMÉ
To reinforce cytotoxic activity and the targeting ability of lymphokine-activated killer cells with a T-cell phenotype (T-LAK) for adoptive immunotherapy against human bile duct carcinoma (BDC), staphylococcal enterotoxin A (SEA) was conjugated chemically with MUSE11 monoclonal antibody (MUSE11 mAb), directed to the MUC1 antigen, using N-succinimidyl 3-(2-pyridyldithio) propionate and 2-iminothiolane HCl. Both SEA-conjugated MUSE11 mAb (SEA-MUSE11) and the F(ab')2 of MUSE11 mAb (SEA-F(ab')2) showed significant enhancement of T-LAK cell tumor neutralization for MUC1 positive-target tumor cells, even with a concentration of 0.01 microg/ml at an E:T ratio of 5:1 in vitro. In this in vitro test, MUC1-positive BDC cells were observed to attach to surrounding T-LAK cells in the presence of SEA-MUSE11 or SEA-F(ab')2. Remarkable tumor growth inhibition was observed in BDC-grafted severe combined immunodeficient mice to which 2 x 10(7) T-LAK cells preincubated with 2 microg of SEA-MUSE11 or SEA-F(ab')2, together with recombinant interleukin 2 (500 IU), were administered i.v. for 4 consecutive days, when tumor size was 5 mm in diameter. These results point to a promising adoptive immunotherapy for patients with BDC.