- Chemistry of single-walled carbon nanotubes.
Chemistry of single-walled carbon nanotubes.
Accounts of chemical research (2002-12-18)
S Niyogi, M A Hamon, H Hu, B Zhao, P Bhowmik, R Sen, M E Itkis, R C Haddon
PMID12484799
RÉSUMÉ
In this Account we highlight the experimental evidence in favor of our view that carbon nanotubes should be considered as a new macromolecular form of carbon with unique properties and with great potential for practical applications. We show that carbon nanotubes may take on properties that are normally associated with molecular species, such as solubility in organic solvents, solution-based chemical transformations, chromatography, and spectroscopy. It is already clear that the nascent field of nanotube chemistry will rival that of the fullerenes.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Carbon nanotube, single-walled, octadecylamine functionalized, 80-90% carbon basis, D × L 2-10 nm × 0.5-2 μm , bundle dimensions, avg. no. of layers, 1