Merck
  • Endothelial glutathione-S-transferase A4-4 protects against oxidative stress and modulates iNOS expression through NF-kappaB translocation.
Endothelial glutathione-S-transferase A4-4 protects against oxidative stress and modulates iNOS expression through NF-kappaB translocation.
Toxicology and applied pharmacology (2008-05-20)
Yongzhen Yang, Yusong Yang, Ya Xu, Scott D Lick, Yogesh C Awasthi, Paul J Boor
RÉSUMÉ
Our recent work in endothelial cells and human atherosclerotic plaque showed that overexpression of glutathione-S-tranferases (GSTs) in endothelium protects against oxidative damage from aldehydes such as 4-HNE. Nuclear factor (NF)-kappaB plays a crucial role during inflammation and immune responses by regulating the expression of inducible genes such as inducible nitric oxide synthase (iNOS). 4-HNE induces apoptosis and affects NF-kappaB mediated gene expression, but conflicting results on 4-HNE's effect on NF-kappaB have been reported. We compared the effect of 4-HNE on iNOS and the NF-kappaB pathway in control mouse pancreatic islet endothelial (MS1) cells and those transfected with mGSTA4, a alpha-class GST with highest activity toward 4-HNE. When treated with 4-HNE, mGSTA4-transfected cells showed significant upregulation of iNOS and nitric oxide (NO) through (NF)-kappaB (p65) translocation in comparison with wild-type or vector-transfected cells. Immunohistochemical studies of early human plaques showed lower 4-HNE content and upregulation of iNOS, which we take to indicate that GSTA4-4 induction acts as an enzymatic defense against high levels of 4-HNE, since 4-HNE accumulated in more advanced plaques, when detoxification and exocytotic mechanisms are likely to be overwhelmed. These studies suggest that GSTA4-4 may play an important defensive role against atherogenesis through detoxification of 4-HNE and upregulation of iNOS.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
GST A4-4, Recombinant Human