Merck
High expression of biglycan is associated with poor prognosis in patients with esophageal squamous cell carcinoma.
International journal of clinical and experimental pathology (2013-11-15)
Ying-Hui Zhu, Fu Yang, Shui-Shen Zhang, Ting-Ting Zeng, Xuan Xie, Xin-Yuan Guan
RÉSUMÉ
Biglycan (BGN), an extracellular matrix component, has been reported to play a crucial role in the tumor progression of various cancers. However, the relation between the expression of BGN and clinical prognosis has not been studied yet. We therefore carry out the present study to elucidate the role of BGN in predicting outcomes of patients with esophageal squamous cell carcinoma (ESCC). In this study, the expression of BGN in 170 cases of ESCC tissues and matched 46 adjacent non-tumorous tissues was measured by quantitative real-time PCR and immunohistochemistry. Upregulation of BGN occurred in approximately 60% of primary ESCCs compared with their non-tumor counterparts. In addition, high expression of BGN was significantly associated with clinical stage (P = 0.009), tumor invasion (P = 0.006) and lymph node metastasis (P = 0.046). The 5-year disease-specific survival (DSS) in high expression of BGN group is poorer than that in low level expression group (36.8% VS 57.4%, P = 0.006). Stratified analysis according to the pathological stage revealed its discernibility on DSS was only pronounced in patients with advanced clinical stage (P = 0.010). Cox multivariate analysis revealed that pathologic N category (P < 0.001; hazard ratio, 2.482, 95% CI, 1.576-3.909) and BGN expression (P = 0.019; hazard ratio, 1.713, 95% CI, 1.092-2.688) were two independent prognostic factors. The findings of the present study provide evidence that BGN represents a potential novel prognostic biomarker for resected ESCC patients in advanced clinical stage.