Merck
  • Selective uptake and imaging of aptamer- and antibody-conjugated hollow nanospheres targeted to epidermal growth factor receptors overexpressed in head and neck cancer.
Selective uptake and imaging of aptamer- and antibody-conjugated hollow nanospheres targeted to epidermal growth factor receptors overexpressed in head and neck cancer.
ACS nano (2014-04-24)
Marites Pasuelo Melancon, Min Zhou, Rui Zhang, Chiyi Xiong, Peter Allen, Xiaoxia Wen, Qian Huang, Michael Wallace, Jeffrey N Myers, R Jason Stafford, Dong Liang, Andrew D Ellington, Chun Li
RÉSUMÉ
The purpose of this study was to compare the binding affinity and selective targeting of aptamer- and antibody-coated hollow gold nanospheres (HAuNS) targeted to epidermal growth factor receptors (EGFR). EGFR-targeting aptamers were conjugated to HAuNS (apt-HAuNS) by attaching a thiol-terminated single-stranded DNA to the HAuNS and then adding the complementary RNA targeted to EGFR. Apt-HAuNS was characterized in terms of size, surface charge, absorption, and number of aptamers per particle. The in vivo pharmacokinetics, in vivo biodistribution, and micro-SPECT/CT imaging of (111)In-labeled apt-HAuNS and anti-EGFR antibody (C225)-conjugated HAuNS were evaluated in nude mice bearing highly malignant human OSC-19 oral tumors. (111)In-labeled PEG-HAuNS was used as a control (n = 5/group). Apt-HAuNS did not have an altered absorbance profile or size (λmax = 800 nm; diameter = 55 nm) compared to C225-HAuNS or PEG-HAuNS. The surface charge became more negative upon conjugation of the aptamer (-51.4 vs -19.0 for PEG-HAuNS and -25.0 for C225-HAuNS). The number of aptamers/particle was ∼250. In vitro cell binding and in vivo biodistribution showed selective binding of the apt-HAuNS to EGFR. μSPECT/CT imaging confirmed that there was more tumor uptake of apt-HAuNS than C225-HAuNS. Aptamer is a promising ligand for image-guided delivery of nanoparticles for treatment of tumor cells overexpressing EGFR.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
EGFR Active human, recombinant, expressed in baculovirus infected insect cells, ≥70% (SDS-PAGE)