Merck
  • Preparation of enantiomerically pure L-7-azatryptophan by an enzymatic method and its application to the development of a fluorimetric activity assay for tryptophanyl-tRNA synthetase.
Preparation of enantiomerically pure L-7-azatryptophan by an enzymatic method and its application to the development of a fluorimetric activity assay for tryptophanyl-tRNA synthetase.
Analytical biochemistry (1997-11-05)
J D Brennan, C W Hogue, B Rajendran, K J Willis, A G Szabo
RÉSUMÉ
The reaction of D,L-7-azatryptophan (D,L-7AW) with tryptophanyl-tRNA synthetase (TrpRS), adenosine triphosphate (ATP), and Mg2+ in the presence of inorganic pyrophosphatase results in the formation of a highly fluorescent l-7AW-adenylate complex. Detection of this complex is based on its enhanced fluorescence at 315 nm excitation and 360 nm emission after the addition of ATP. This stereoselective reaction was used to develop an activity assay for TrpRS using commercially available racemic D,L-7AW. The assay can be used to determine the activity of TrpRS from samples which contain less than 1 nmol of enzyme in 250 microL of sample. Thus the enzyme activity can be assessed without resorting to a radioactive assay of tRNATrp acylation. A secondary use of the stereoselective assay was for confirming the presence of pure L-7AW, D-7AW, or mixtures of the two enantiomers. D-7AW and L-7AW were prepared by reacting D,L-7AW with chloroacetic anhydride to form N-chloroacetyl-D,L-7AW (ClAc-7AW) followed by stereospecific proteolytic digestion of ClAc-7AW using carboxypeptidase A to produce the free L-7AW. The L-7AW could be separated from unreacted N-chloroacetyl-7AW by reverse-phase HPLC. The TrpRS-based assay was able to unambiguously discriminate between the two enantiomers of 7AW. The assay was then used to identify which enantiomer of 7AW was present in resolved fractions of the tripeptide L-lysyl-D,L-7-azatryptophyl-L-lysine. Digestion of the resolved tripeptides with protease enzymes produced the free L or D enantiomer of 7AW, which was easily identified using the TrpRS assay procedure.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Chloroacetic anhydride, 95%