Merck
Enhanced production of ansamitocin P-3 by addition of Mg2+ in fermentation of Actinosynnema pretiosum.
Bioresource technology (2011-09-13)
Yongliang Jia, Jian-Jiang Zhong
RÉSUMÉ
The effect of divalent metal ions (i.e., Mn(2+), Mg(2+), Zn(2+), Cu(2+), and Co(2+)) on the production of anticancer ansamitocin P-3 (AP-3) by submerged cultures of Actinosynnema pretiosum in medium containing agro-industrial residues was investigated, and Mg(2+) was found to be the most effective. Under the optimal condition of Mg(2+) addition, the maximal AP-3 production titer reached 85 mg/L, which was 3.0-fold that of the control. The activities of methylmalonyl-CoA carboxyltransferase (MCT) and methylmalonyl-CoA mutase (MCM) were enhanced. The content of two precursors, malonyl-CoA and methylmalonyl-CoA, was lower than that of control. This work demonstrates that Mg(2+) addition is a simple and effective strategy for increasing AP-3 production through the regulation of enzyme activity and pools of precursors. The information obtained can be helpful to its efficient production on large scale.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Ansamitocin P-3 from Actinosynnema pretiosum, ≥90% (HPLC)