Merck
A phase I trial of lomeguatrib and irinotecan in metastatic colorectal cancer.
Cancer chemotherapy and pharmacology (2009-12-30)
A Sabharwal, P G Corrie, R S Midgley, C Palmer, J Brady, P Mortimer, A J Watson, G P Margison, M R Middleton
RÉSUMÉ
Expression of the DNA repair protein O (6)-methylguanine-DNA methyltransferase (MGMT) correlates with resistance to irinotecan in colorectal cancer cell lines. This phase I study evaluated the maximum tolerated dose (MTD) of lomeguatrib, an inactivating pseudosubstrate of MGMT, in combination with irinotecan in patients with metastatic colorectal cancer and assessed the safety, toxicity and clinical pharmacology of combination treatment. Patients with metastatic colorectal cancer received lomeguatrib (10-80 mg PO) on days 1-5 with irinotecan (250-350 mg/m(2) IV) on day 4 of a 21-day cycle. Twenty-four patients, pre-treated with a median of 2 lines of chemotherapy, received 104 cycles of treatment. The MTD was defined as 80 mg/day lomeguatrib with 300 mg/m(2) irinotecan. The main toxicities observed were neutropaenia and diarrhoea. Lomeguatrib of 80 mg/day produced complete MGMT depletion in all available peripheral blood mononuclear cells (PBMCs) and paired tumour biopsies (one patient). There was no pharmacokinetic interaction between the drugs. In 22 patients assessable for tumour response, one achieved a partial response and 16 had stable disease. This study defined a tolerable dose of irinotecan in combination with lomeguatrib in patients with metastatic colorectal cancer. Combination treatment gave a similar response rate to irinotecan monotherapy in this heavily pre-treated patient group.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Lomeguatrib, ≥98% (HPLC)