Merck
The plasma pharmacokinetics of iophenoxic and iopanoic acids in goat.
Xenobiotica; the fate of foreign compounds in biological systems (1992-02-01)
C T Eason, C M Frampton
RÉSUMÉ
1. The comparative plasma pharmacokinetics of two organic iodine-containing compounds were evaluated in the goat for their suitability as markers in wildlife studies. 2. After oral administration of a single dose, the plasma elimination half-life for iopanoic acid was considerably more rapid (t1/2 of 1-2 days) than that of iophenoxic acid (t1/2 of 81 days). 3. Similar peak plasma concentrations were obtained after administration of iophenoxic acid (1.5 mg/kg) and iopanoic acid (25 mg/kg); however, the AUC0----infinity for iopanoic acid at doses of 25, 50, and 100 mg/kg were 201 +/- 39, 604 +/- 225, and 1292 +/- 721 (micrograms h/ml +/- SD), respectively, which were less than the value of 36,600 +/- 6387 for the oral administration of iophenoxic acid at 1.5 mg/kg. 4. Iophenoxic acid was chosen as a suitable marker because of its persistence at detectable concentrations in the plasma for 5 months.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
α-Ethyl-3-hydroxy-2,4,6-triiodohydrocinnamic acid, 97%