Merck
Synthesis of 2-(trimethylsilyl)ethyloxycarbonylamino acids and their use in peptide chemistry.
Hoppe-Seyler's Zeitschrift fur physiologische Chemie (1981-09-01)
B Wünsch, L Moroder, O Keller
RÉSUMÉ
2-(Trimethylsilyl)ethyl-4-nitrophenyl carbonate has been prepared as a new reagent for the introduction of the 2-(trimethylsilyl)ethyloxycarbonyl group into amino acids or amino acid derivatives. The resulting N alpha-protected amino acids were found to represent suitable intermediates for the synthesis of peptides. The amino-protecting group proved to be stable under the usual conditions of peptide synthesis and readily cleaved in selective conditions via fluoride ion-mediated fragmentation.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
4-Nitrophenyl 2-(trimethylsilyl)ethyl carbonate, ≥97.0%