Merck
High-performance liquid chromatography-electrochemical detection of 3-methylhistidine in human urine.
Journal of chromatography (1992-12-23)
P Betto, G Ricciarello, S Pichini, L Dello Strologo, G Rizzoni
RÉSUMÉ
A reversed-phase high-performance liquid chromatographic method is described for the determination of 3-methylhistidine content in human urine using pre-column derivatization with phenylisothiocyanate, isocratic elution with 15 mM sodium acetate-acetonitrile (92:8, v/v) and electrochemical detection. The limit of quantitation was 0.1 pmol. The method has been applied in routine analyses of 3-methylhistidine in both clinical and research work.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1-Methyl-L-histidine, ≥98.0% (TLC)