Merck
Benzoic acid metabolism in presence of Schistosoma mansoni infection in mice.
Indian journal of experimental biology (1989-02-01)
T S Emundianughe
RÉSUMÉ
The metabolism of benzoic acid was examined in S. mansoni infected CBA mouse. The result showed that control animals dosed with 150 mg/kg benzoic acid resulted in urinary excretion of two metabolites, hippuric acid and benzoic acid glucuronide. Administration of the same dose to animal carrying S. mansoni for a period of over 6 weeks resulted in decreased formation of hippuric acid and total elimination of benzoic acid by glucuronide pathway.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Transglutaminase from guinea pig liver, ≥1.5 units/mg protein, recombinant, expressed in E. coli