Merck
Diminished cell proliferation promotes natural killer cell adaptive-like phenotype by limiting FcεRIγ expression.
The Journal of experimental medicine (2022-09-07)
Avishai Shemesh, Yapeng Su, Daniel R Calabrese, Daniel Chen, Janice Arakawa-Hoyt, Kole T Roybal, James R Heath, John R Greenland, Lewis L Lanier
RÉSUMÉ
Human adaptive-like natural killer (NK) cells express low levels of FcεRIγ (FcRγ-/low) and are reported to accumulate during COVID-19 infection; however, the mechanism underlying and regulating FcRγ expression in NK cells has yet to be fully defined. We observed lower FcRγ protein expression in NK cell subsets from lung transplant patients during rapamycin treatment, suggesting a link with reduced mTOR activity. Further, FcRγ-/low NK cell subsets from healthy donors displayed reduced mTOR activity. We discovered that FcRγ upregulation is dependent on cell proliferation progression mediated by IL-2, IL-15, or IL-12, is sensitive to mTOR suppression, and is inhibited by TGFβ or IFNα. Accordingly, the accumulation of adaptive-like FcRγ-/low NK cells in COVID-19 patients corresponded to increased TGFβ and IFNα levels and disease severity. Our results show that an adaptive-like NK cell phenotype is induced by diminished cell proliferation and has an early prognostic value for increased TGFβ and IFNα levels in COVID-19 infection associated with disease severity.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Rapamycin, ≥95% (HPLC), powder
Sigma-Aldrich
Anticorps Milli-Mark® anti-FcεRI, sous-unité γ, conjugué FITC, Milli-Mark®, from rabbit
Sigma-Aldrich
Foxo1 Inhibitor, AS1842856, Foxo1 Inhibitor, AS1842856, is a cell-permeable inhibitor that blocks the transcription activity of Foxo1 (IC₅₀ = 33 nM). Directly binds to the active Foxo1, but not the Ser256-phosphorylated form.