Merck
Lithium iodide as a promising electrolyte additive for lithium-sulfur batteries: mechanisms of performance enhancement.
Advanced materials (Deerfield Beach, Fla.) (2014-11-05)
Feixiang Wu, Jung Tae Lee, Naoki Nitta, Hyea Kim, Oleg Borodin, Gleb Yushin
RÉSUMÉ
Lithium Iodide (LiI) is reported as a promising electrolyte additive for lithium-sulfur batteries. It induces formation of Li-ion-permeable protective coatings on both positive and negative electrodes, which prevent the dissolution of polysulfides on the cathode and reduction of polysulfides on the anode. In addition to enhancing the cell cycle stability, LiI addition also decreases the cell overpotential and voltage hysteresis.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Lithium iodide, 99.9% trace metals basis