Merck
IL-18, although antiallergic when administered with IL-12, stimulates IL-4 and histamine release by basophils.
Proceedings of the National Academy of Sciences of the United States of America (1999-11-26)
T Yoshimoto, H Tsutsui, K Tominaga, K Hoshino, H Okamura, S Akira, W E Paul, K Nakanishi
RÉSUMÉ
IL-18 is a proinflammatory cytokine that plays an important role in natural killer cell activation and T helper 1 (Th1) cell responses. Mast cells and basophils are major inducers and effectors of allergic inflammation. Here we show that basophils and mast cells derived by culture of bone marrow cells with IL-3 for 10 days express IL-18Ralpha chain and that basophils produce large amounts of IL-4 and IL-13 in response to stimulation with IL-3 and IL-18. Injection of IL-12 and IL-18 inhibits IgE production in helminth-infected wild-type mice and abolishes the capacity of their basophils to produce IL-4 and IL-13 in response to stimulation either with IL-3 and IL-18 or with FcepsilonR cross-linkage. By contrast, this combination of cytokines actually increases IgE levels in helminth-infected IFN-gamma(-/-) mice and enhances IL-4 and IL-13 production by their basophils. Furthermore, injection of IL-18 alone enhances basophil production of IL-4 and histamine both in wild-type and IFN-gamma(-/-) mice. Thus, IL-18 has the potential to stimulate basophils but, when given with IL-12, exhibits an antiallergic action in vivo.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Mouse Il18 / Interleukin-18 ELISA Kit, for serum, plasma and cell culture supernatants