Merck
Carbonic anhydrase inhibitors: interactions of phenols with the 12 catalytically active mammalian isoforms (CA I-XIV).
Bioorganic & medicinal chemistry letters (2008-02-05)
Alessio Innocenti, Daniela Vullo, Andrea Scozzafava, Claudiu T Supuran
RÉSUMÉ
The inhibition of the metalloenzyme carbonic anhydrase (CA, EC 4.2.1.1) with three phenols was investigated. Phenol was an effective CA I-IV, IX, XII and XIV inhibitor (K(I)s of 2.7-11.5 microM) and a less effective one against the other isoforms, CA VA, VB, VI, VII, and XIII (K(I)s of 208-710 micraoM). 3,5-Difluorophenol was an effective inhibitor of CA III, IV, IX, and XIV (K(I)s of 0.71-10.7 microM) being a weaker one for CA I, II, VA, VB, VI, VII, XII, and XIII (K(I)s of 33.9-163 microM). Clioquinol (5-chloro-7-iodo-8-quinolinol) was the best phenol inhibitor against all isozymes, with inhibition constants in the range of 3.3-16.0 microM. These data prove that the phenol OH moiety can be considered as a new 'zinc-water binding group' for the design of CA inhibitors possessing a different inhibition mechanism as compared to the classical sulfonamide inhibitors that bind the metal ion within the active site cavity.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
5-Chloro-7-iodo-8-quinolinol, ≥95.0% (HPLC)
Sigma-Aldrich
3,5-Difluorophenol, 99%