Merck
Phospholipid profiles of control and glaucomatous human aqueous humor.
Biochimie (2014-02-25)
Genea Edwards, Katyayini Aribindi, Yenifer Guerra, Richard K Lee, Sanjoy K Bhattacharya
RÉSUMÉ
To compare phospholipid (phosphatidylcholine, phosphatidylserine, phosphatidylethanolamine and phosphatidylinositol) profiles of human control and glaucomatous aqueous humor (AQH). AQH samples were procured during surgery from human POAG and control subjects (n = 15 each). Samples were used following institutional review board approved protocols and adhering to the tenets of the Declaration of Helsinki. Lipid extraction was performed using a modification of the Bligh and Dyer method, protein concentrations were determined using the Bradford's method, and select samples were confirmed with Densitometry of PHAST gels. Lipids were identified and subjected to ratiometric quantification using a TSQ Quantum Access Max triple quadrupole mass spectrometer utilizing precursor ion scan (PIS) or neutral ion loss scan (NLS) using appropriate class specific lipid standards in a two step quantification process. The comparative profiles of phosphatidylcholines, phosphatidylserines, phosphatidylethanolamines and phosphatidylinositols between control and glaucomatous AQH showed several species common between them. A number of unique lipids in all four phospholipid classes were also identified in control eyes that were absent in glaucomatous eyes and vice versa. A number of phospholipids were found to be uniquely present in control, but absent in glaucomatous AQH and vice versa. Compared with a previous study of control and POAG red blood cells, a number of these phospholipids are absent locally (AQH).
MATÉRIAUX
Référence du produit
Marque
Description du produit
Avanti
13:0 PC, 1,2-ditridecanoyl-sn-glycero-3-phosphocholine, chloroform