Merck
  • Derivation of a human induced pluripotent stem cell line (QBRIi007-A) from a patient carrying a homozygous intronic mutation (c.613-7T>G) in the SLC2A2 gene.
Derivation of a human induced pluripotent stem cell line (QBRIi007-A) from a patient carrying a homozygous intronic mutation (c.613-7T>G) in the SLC2A2 gene.
Stem cell research (2020-03-09)
Ahmed K Elsayed, Maryam Aghadi, Sara Al-Khawaga, Khalid Hussain, Essam M Abdelalim
RÉSUMÉ
Fanconi Bickel Syndrome (FBS) is an autosomal recessive disease resulting from mutations in the SLC2A2 gene, encoding the GLUT2. FBS patients develop diabetes mellitus. Using non-integrating Sendai virus, we generated an induced pluripotent stem cell (iPSC) line, QBRIi007-A, carrying the c.613-7 T>G homozygous mutation in intron 5 of the SLC2A2 gene from a 19-year-old female with FBS and diabetes. The iPSC line was characterized for pluripotency, differentiation potential, genomic integrity, and genetic identity. This iPSC line provides a useful cell model to understand the role of GLUT2 in the disease development and to discover new drug candidates.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Kit de détection de la phosphatase alcaline, This Alkaline Phosphatase Detection Kit is a specific & sensitive tool for the phenotypic assessment of Embryonic Stem (ES) cell differentiation by the determination of AP activity.
Sigma-Aldrich
Triton X-100, laboratory grade