Merck
Activation of protein kinase G up-regulates expression of 15-lipoxygenase-1 in human colon cancer cells.
Cancer research (2005-09-17)
Atsuko Deguchi, Steven W Xing, Imad Shureiqi, Peiying Yang, Robert A Newman, Scott M Lippman, Steven J Feinmark, Bert Oehlen, I Bernard Weinstein
RÉSUMÉ
Recent studies indicate that the induction of apoptosis in human colon cancer cells by certain nonsteroidal antiinflammatory drugs involves increased expression of 15-LOX-1 and synthesis of its major product 13-S-hydroxyoctadecadienoic acid (13-S-HODE). Evidence was obtained that this occurs via a cyclooxygenase-2 (COX-2)-independent mechanism, but the actual mechanism of induction of 15-LOX-1 by these compounds is not known. There is extensive evidence that treatment of SW480 human colon cancer cells with sulindac sulfone (Exisulind, Aptosyn) or the related derivative OSI-461, both of which inhibit cyclic GMP (cGMP)-phosphodiesterases but lack COX-2 inhibitory activity, causes an increase in intracellular levels of cGMP, thus activating protein kinase G (PKG), which then activates pathways that lead to apoptosis. Therefore, in the present study, we examined the effects of various agents that cause increased cellular levels of cGMP on the expression of 15-LOX-1 in SW480 human colon cancer cells. Treatment of the cells with Exisulind, sulindac sulfide, OSI-461, the guanylyl cyclase activator YC-1, or the cell-permeable cGMP compound 8-para-chlorophenylthio-cGMP (8-pCPT-cGMP) caused an increase in cellular levels of 15-LOX-1. Exisulind, OSI-461, and 8-pCPT-cGMP also increased mRNA levels of 15-LOX-1, suggesting that the effects were at the level of transcription. The cGMP-phosphodiesterase inhibitors and YC-1 increased the production of 13-S-HODE, which is the linoleic acid metabolite of 15-LOX-1. Treatment of SW480 cells with the PKG inhibitor Rp-8-pCPT-cGMP blocked Exisulind-induced 15-LOX-1 expression. Furthermore, derivatives of SW480 cells that were engineered to stably overexpress wild-type PKG Ibeta displayed increased cellular levels of 15-LOX-1 when compared with vector control cells. Taken together, these results provide evidence that the cGMP/PKG pathway can play an important role in the induction of 15-LOX-1 expression by nonsteroidal antiinflammatory drugs and related agents.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Protein Kinase G Iβ human, ≥95% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf9 cells, buffered aqueous glycerol solution