Merck
  • Ruminal fermentation, kinetics, and total-tract digestibility of lactating dairy cows fed distillers dried grains with solubles in low- and high-forage diets.
Ruminal fermentation, kinetics, and total-tract digestibility of lactating dairy cows fed distillers dried grains with solubles in low- and high-forage diets.
Journal of dairy science (2019-07-01)
Sanjeewa D Ranathunga, Kenneth F Kalscheur, Kevin J Herrick
RÉSUMÉ
The objective of this study was to investigate the effects of concentrations of forages and corn distillers dried grains with solubles (DDGS) on ruminal fermentation, ruminal kinetics, and total-tract digestibility of nutrients in lactating dairy cows. Four lactating Holstein cows with ruminal cannulas were assigned to a 4 × 4 Latin square in a 2 × 2 factorial arrangement of treatments. Diets were formulated to contain low forage [LF; 17% forage neutral detergent fiber (NDF)] or high forage (HF; 24.5% forage NDF) and DDGS at 0 or 18% (0DG or 18DG) of diet dry matter (DM). Intake of DM was not affected by the diets. Daily mean ruminal pH was affected by forage NDF × DDGS interactions, as the lowest ruminal pH was observed among cows fed LF18DG (6.02). Apparent total-tract digestibility for DM, organic matter, crude protein, NDF, acid detergent fiber, and starch was not affected by diets. Cows fed LF diets had a greater total volatile fatty acid concentration compared with cows fed HF (122 vs. 116 mM). Molar proportions of acetate were greater for HF compared with that of LF diets (62.6 vs. 57.5 mmol/100 mmol) and greater for 0DG diet compared with that of 18DG diets (61.3 vs. 58.7 mmol/100 mmol) diets. The molar proportion of propionate was affected by forage × DDGS interaction as the greatest propionate molar proportion was observed with cows fed LF18DG diet (27.7 mmol/100 mmol). Also, molar proportion of butyrate was affected by forage × DDGS interaction, as the greatest butyrate molar proportion was observed in cows fed HF18DG diet (13.5 mmol/100 mmol). Average fractional dilution rate for all diets was 11.9%/h and was not affected by diets. Fractional passage rate of the solid phase was greater for HF than for LF (4.40 vs. 3.76%/h). The ruminal retention time of solid phase was greater for LF compared with HF diets (27.3 vs. 23.3 h). Fractional passage rate of DDGS was affected by forage × DG interaction, as the highest fractional passage rate of DDGS was observed with cows fed HF18DG diet (7.72%/h). Our results demonstrated that concentrations of forage, DDGS, and their interaction influence ruminal degradation and kinetics of diets fed. Diets formulated at 17% forage NDF at 17% (DM basis) can decrease milk fat concentration compared with diets formulated at 25% forage NDF. Additionally, feeding DDGS at 18% DM basis to lactating dairy cows did not affect milk fat concentration or yield.