Merck
Effect of Feeding Pomegranate Byproduct on Fatty Acid Composition of Ruminal Digesta, Liver, and Muscle in Lambs.
Journal of agricultural and food chemistry (2019-04-02)
Antonio Natalello, Giuseppe Luciano, Luciano Morbidini, Bernardo Valenti, Mariano Pauselli, Pilar Frutos, Luisa Biondi, Pablo J Rufino-Moya, Massimiliano Lanza, Alessandro Priolo
RÉSUMÉ
This work investigated the effects of feeding whole pomegranate byproduct (WPB) to lambs on ruminal, liver, and intramuscular fatty acids (FA). Seventeen lambs, divided into two groups, were fed for 36 days with a cereal-based concentrate diet (CON) or with a concentrate diet containing 200 g/kg DM of WPB to partially replace barley and corn (WPB). The dietary treatment did not affect the final body and carcass weight, the dry matter intake, or the average daily gain. However, total polyunsaturated FA (PUFA), linolenic, rumenic (RA), and vaccenic (VA) acid were increased in liver (+15%, +32%, +344%, and +118%, respectively) and muscle (+46%, +38%, +169%, and +89%, respectively) of WPB lambs ( P < 0.05). Punicic acid and three isomers of conjugated linolenic acid were detected exclusively in the rumen and tissues of WPB-lambs. The C18:1 t10/ t11 ra tio in rumen digesta or in tissues was reduced by feeding WPB (-791%, -690%, and -456%, respectively, in rumen, liver and muscle; P < 0.001), suggesting that the WPB prevented the t10-shift rumen biohydrogenation pathway. In conclusion, the inclusion of WPB into a concentrate-based diet can be a strategy to improve the FA composition of meat, without effects on the animal performances.