Merck
  • Formation and Disordered Degradation of Neutrophil Extracellular Traps in Necrotizing Lesions of Anti-Neutrophil Cytoplasmic Antibody-Associated Vasculitis.
Formation and Disordered Degradation of Neutrophil Extracellular Traps in Necrotizing Lesions of Anti-Neutrophil Cytoplasmic Antibody-Associated Vasculitis.
The American journal of pathology (2019-01-25)
Sakiko Masuda, Mayu Nonokawa, Emika Futamata, Yuka Nishibata, Sari Iwasaki, Takahiro Tsuji, Yutaka Hatanaka, Daigo Nakazawa, Satoshi Tanaka, Utano Tomaru, Tamihiro Kawakami, Tatsuya Atsumi, Akihiro Ishizu
RÉSUMÉ
Antineutrophil cytoplasmic antibody (ANCA)-associated vasculitis (AAV) is characterized by the production of ANCAs and systemic necrotizing vasculitis in small vessels. Disordered regulation of neutrophil extracellular traps (NETs) is critically involved in the pathogenesis of AAV. NETs are web-like DNA decorated with antimicrobial proteins; they are extruded from activated neutrophils. The principal degradation factor of NETs in vivo is DNase I; however, NETs resistant to DNase I can persist in tissues and can lead to the production of ANCAs. Deposition of NETs has been demonstrated in glomerular crescents and necrotizing vasculitis in AAV. Here, the amount of NETs in formalin-fixed, paraffin-embedded tissue sections was examined, and the results for AAV were compared with the results for diseases that should be distinguished from AAV. NETs were more abundant in necrotizing vasculitis of AAV than in non-ANCA-associated vasculitis, or in granulomatous angiitis. Pulmonary granulomas in AAV and non-ANCA-associated diseases were further studied. The amount of NETs was significantly greater in necrotizing granulomas of AAV than in granulomas of sarcoidosis without necrosis. Although NETs were formed in necrotizing granulomas of tuberculosis equivalently to those formed in AAV, they were more susceptible to degradation by DNase I than were NETs in AAV. The formation and disordered degradation of NETs in necrotizing lesions are characteristics of AAV and are possibly related to its pathogenesis.