Merck
  • Determination of twenty pharmaceutical contaminants in soil using ultrasound-assisted extraction with gas chromatography-mass spectrometric detection.
Determination of twenty pharmaceutical contaminants in soil using ultrasound-assisted extraction with gas chromatography-mass spectrometric detection.
Chemosphere (2019-06-04)
Jolanta Kumirska, Paulina Łukaszewicz, Magda Caban, Natalia Migowska, Alina Plenis, Anna Białk-Bielińska, Małgorzata Czerwicka, Fei Qi, Stepnowski Piotr
RÉSUMÉ
In this paper, an analytical method for the simultaneous determination of twenty pharmaceuticals (eight non-steroidal anti-inflammatory drugs, five oestrogenic hormones, two antiepileptic drugs, two β-blockers, and three antidepressants) in soils was developed. The optimal method included ultrasound-assisted extraction, a clean-up step on a silica gel column, derivatization using N,O-bis(trimethylsilyl)trifluoroacetamide (BSTFA) and 1% trimethylchlorosilane (TMCS) in pyridine and ethyl acetate (2:1:1, v/v/v) for 30 min at 60 °C, and determination by gas chromatography-mass spectrometry working in the selected ion monitoring mode. This affords good resolution, high sensitivity and reproducibility, and freedom from interferences even from complex matrices such as soils. The method detection limits ranged from 0.3 to 1.7 ng g-1, the intra-day precision represented as RSDs ranged from 1.1 to 10.0%, and the intra-day accuracy from 81.3 to 119.7%. The absolute recoveries of the target compounds were above 80%, except for valproic acid and diethylstilbestrol. The developed method was successfully applied in the analysis of the target compounds in soils collected in Poland. Among the 20 pharmaceuticals, 12 compounds were detected at least once in the soils. The determination of antiepileptic drugs, β-blockers, and antidepressants was also performed for the first time.