Merck
TDP-43 Neuropathologic Associations in the Nun Study and the Honolulu-Asia Aging Study.
Journal of Alzheimer's disease : JAD (2018-11-20)
Margaret E Flanagan, Brenna Cholerton, Caitlin S Latimer, Laura S Hemmy, Steven D Edland, Kathleen S Montine, Lon R White, Thomas J Montine
RÉSUMÉ
Transactive response binding protein-43 (TDP-43) cytoplasmic neuronal and glial aggregates (pathologic TDP-43) have been described in multiple brain diseases. We describe the associations between neuropathologically confirmed TDP-43 and cognition in two population-based cohorts: the Nun Study (NS) and the Honolulu-Asia Aging Study (HAAS). In the HAAS, there was a significant association between hippocampal sclerosis (HS) and TDP-43 (OR = 11.04, p < 0.0001, 95% CI 3.57-34.13). In the NS, there were significant associations between TDP-43 and HS (OR = 16.44, p > 0.001 95%, CI 7.10-38.00) and Alzheimer's disease (AD) severity (OR = 1.74, p = 0.009, 95% CI 1.15-2.64). When cognitive scores were added to the model, HS remained significant but the other variables were not. When HS was removed from the model, the overall model remained significant and the associations between cognitive performance and TDP-43 (OR = 2.11, p = 0.022, 95% CI 1.11-4.02) were significant. In the NS, there was a significant association between cognitive performance and TDP-43 (OR 1.94 p = 0.005, 95% CI 1.22-3.09) (HS remained significant, but AD did not). When HS was removed from the model, only CERAD was significant (OR = 2.43 p < 0.001, 95% CI 1.58-3.74). These results support a consistent association between pathologic TDP-43, HS, and the development of cognitive impairment in two large studies of brain aging, while the relationship between AD pathology and TDP-43 may vary according to cohort-specific features.