Merck
Purification and Nitrogen Doping of Nanothin Exfoliated Graphite Through RF Thermal Plasma Treatment.
Nanomaterials (Basel, Switzerland) (2019-07-13)
Byung-Koo Son, Kyu-Hang Lee, Tae-Hee Kim, Myung-Sun Shin, Sun-Yong Choi, Guangsup Cho
RÉSUMÉ
A mixture of nanothin exfoliated (NTE) graphite and urea (CO(NH2)2) powder was treated with radio frequency (RF) thermal plasma to achieve in situ purification and nitrogen doping of NTE graphite using the high-temperature flame of the RF plasma. Reactive species such as NH3, NH2, and HCNO generated by the thermolysis of urea play an important role in the purification and nitrogen doping of NTE graphite. The nitrogen content of NTE graphite subjected to plasma treatment increased by 5 times compared with that of raw NTE graphite. Three types of nitrogen species, namely, quaternary N, pyridinic N, and pyrrolic N, were observed after N doping with plasma treatment. The sheet resistance of N-doped NTE graphite reduced to 12-21% compared to that of the untreated NTE graphite, with the corresponding resistivity being ~7 × 10-6 Ω m.