Merck
Linezolid Inhibited Synthesis of ATP in Mitochondria: Based on GC-MS Metabolomics and HPLC Method.
BioMed research international (2018-11-10)
Xuemei Ye, Aifang Huang, Xianqin Wang, Congcong Wen, Lufeng Hu, Guanyang Lin
RÉSUMÉ
Linezolid has been widely used in serious infections for its effective inhibiting effect against multidrug-resistant gram-positive pathogens. However, linezolid caused severe adverse reactions, such as thrombocytopenia, anaemia, optic neuropathy, and near-fatal serotonin syndrome. In order to investigate the toxicity of linezolid, twenty-four Sprague-Dawley rats were randomly divided into: control group (n=7), low-group (n=8), and high-group (n=9). The rats of low-group and high-group were given by gavage with linezolid 60 and 120 mg/kg/day for 7 days, respectively. The serum concentration of linezolid was determined by high performance liquid chromatography (HPLC); blood metabolic change was analyzed by gas chromatography-mass spectrometer (GC-MS). Adenosine triphosphate (ATP) concentration in HepG2-C3A after being cultured with linezolid was determined by HPLC. The results showed that there were six metabolites and nine metabolites had statistical differences in low-group and high-group (P<0.05). The trimethyl phosphate was the most significant indicator in those changed metabolites. Except for d-glucose which was slightly increased in low-group, octadecanoic acid, cholest-5-ene, hexadecanoic acid, α-linolenic acid, eicosapentaenoic acid, 9,12-Octadecadienoic acid, and docosahexaenoic acid were all decreased in low-group and high-group. ATP concentration was decreased in HepG2-C3A after cultured with linezolid. In conclusion, the toxicity of linezolid is related to its serum concentration. Linezolid may inhibit the synthesis of ATP and fatty acid.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
N-Methylhydroxylamine hydrochloride, 98%