Key Documents
GBL400104
Grace Bio-Labs slide imaging tray
wells, 4, For 25 mm X 75 mm microscope slides
Synonyme(s) :
multi slide viewer, slide viewer
About This Item
Code UNSPSC :
41122600
Nomenclature NACRES :
NB.17
Caractéristiques
wells 4
For 25 mm X 75 mm microscope slides
Fabricant/nom de marque
Grace Bio-Labs 400104
Description générale
Imaging Tray to hold 4 microscope slides, slide dimensions 25mm X 75mm.