Key Documents
Z406295
Sigma-Aldrich
Sure/Seal bottles
capacity 125 mL, amber plastic-coated glass
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux
amber plastic-coated glass
round bottle
Joint
joint threaded neck
Caractéristiques
without cap
Conditionnement
pkg of 6 ea
Fabricant/nom de marque
Sigma-Aldrich
Capacité
125 mL
Description générale
Order overcaps, steel crown caps, and PTFE-faced liners separately.
Application
Bottles are used to store chemicals, solvents, and other liquids. Amber-tinted glass is recommended for light-sensitive materials.
Informations légales
Sure/Seal is a trademark of Sigma-Aldrich Co. LLC