Key Documents
Z319449
Büchi® evaporating flask, pear-shaped
glass, volume 250 mL, joint: ST/NS 29/32
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
glass
pear-shaped
single-neck
Joint
joint: ST/NS29/32
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Büchi 00433
Capacité
250 mL
Volume
250 mL
Description générale
These flasks fit R200/205, R210/215, and RII evaporators
Informations légales
Büchi is a registered trademark of Büchi Labortechnik AG