Key Documents
Z317403
Duran® desiccator
I.D. 100 mm
Synonyme(s) :
247814603
About This Item
Code UNSPSC :
41104510
Nomenclature NACRES :
NB.43
Fabricant/nom de marque
DURAN®
Schott
Disponibilité
not available in USA and Canada
Hauteur
187 mm
D.I.
100 mm
Description générale
Flat flange, and cover with knob.
Informations légales
DURAN is a registered trademark of DWK Life Sciences