Z249602
Aldrich® modified, jacketed Dean-Stark trap
joint: ST/NS 24/40, Overall H 300 mm
About This Item
Produits recommandés
Joint
joint: ST/NS24/40
H totale
300 mm
Description générale
Designed specifically for azeotropic distillation using solvents heavier than water (e.g., chloroform). 350 mL capacity. 0-3 mm PTFE valve.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC