Key Documents
Z215899
Solid PTFE stopper
joint: ST/NS 10/30
Synonyme(s) :
flask stopper, lab stopper, stoppers
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.35
Joint
joint: ST/NS10/30
Fabricant/nom de marque
Cowie 010.110
Description générale
Machined from solid PTFE with trapezoidal seals for medium to high vacuum performance up to 280°C (540°F).