TEM-CF300NI
Carbon Film Supported Nickel Square Mesh
size 300 mesh, box of 25
Synonyme(s) :
CARBON FILM 300# NICKEL 25/PK
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Produits recommandés
Conditionnement
box of 25
Taille
300 mesh
Description générale
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Nickel grid, 300 mesh.