Key Documents
HAM59000-50
Hamilton® Microlab® 600 instrument syringe
model, 1010 BFP, volume 10 mL
Synonyme(s) :
Hamilton syringe
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
Matériaux
Gastight Borosilicate glass/PTFE syringe
Caractéristiques
model 1010 BFP
Fabricant/nom de marque
Hamilton 59000-50
Paramètres
10-115 °C temp. range (50-239 °F)
200 psig pressure
D.I. × D.E.
14.6 mm × 17.7 mm
Volume
10 mL
Informations légales
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.