Key Documents
BR708709
BRAND® disposable BLAUBRAND® micropipettes, intraMark
tube capacity 10 μL, orange
Synonyme(s) :
Micropipettes, Pipettes, intraMARK
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
Matériaux
orange
Conditionnement
pack of 1000 ea (4 boxes of 250)
Fabricant/nom de marque
BRAND 708709
Longueur
125 mm
Capacité du tube
10 μL
Description générale
DIN ISO 7550. DE-M marking for volumes greater than 5 µl. Calibrated to contain (TC, In). Color code to ISO for clear identification.
Informations légales
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG