Merck
  • Sphingolipids are essential for the growth of Chinese hamster ovary cells. Restoration of the growth of a mutant defective in sphingoid base biosynthesis by exogenous sphingolipids.
Sphingolipids are essential for the growth of Chinese hamster ovary cells. Restoration of the growth of a mutant defective in sphingoid base biosynthesis by exogenous sphingolipids.
The Journal of biological chemistry (1992-11-25)
K Hanada, M Nishijima, M Kiso, A Hasegawa, S Fujita, T Ogawa, Y Akamatsu
ABSTRACT
We previously isolated a temperature-sensitive Chinese hamster ovary cell mutant (strain SPB-1) with thermolabile serine palmitoyltransferase, which is involved in the first step of sphingolipid synthesis (Hanada, K., Nishijima, M., and Akamatsu, Y. (1990) J. Biol. Chem. 265, 22137-22142). In this study, sphingolipid-deficient culture medium was used to examine the effect of exogenous sphingolipids on the cell growth of SPB-1. When cultivated in the sphingolipid-deficient medium, SPB-1 cells ceased growing at non-permissive temperatures. Under these conditions, de novo sphingolipid synthesis ceased in the SPB-1 cells, resulting in a decrease in levels of sphingomyelin and ganglioside sialyl lactosylceramide (GM3), whereas the parental CHO-K1 cells grew logarithmically with normal sphingolipid synthesis. Exogenous sphingosine restored the contents of both sphingomyelin and GM3 in the SPB-1 cells near to the parental levels through metabolic utilization and allowed the mutant cells to grow even at the non-permissive temperature. Similarly, exogenous sphingomyelin restored the sphingomyelin levels and only partly the GM3 levels and also suppressed the temperature-sensitivity of the SPB-1 cell growth. In contrast, exogenous glucosylceramide, which restored the GM3 levels but not the sphingomyelin levels, failed to suppress the temperature sensitivity of the SPB-1 cell growth. Combination of exogenous sphingomyelin with ceramide, glucosylceramide, GM3, or sphingoid bases did not show any synergistic or additive effect on the SPB-1 cell growth enhancement, compared with sphingomyelin alone. The results indicated that the temperature sensitivity of the SPB-1 cell growth was due to the lack of cellular sphingolipids, possibly that of sphingomyelin.
Product Description
Supelco
SPB®-1 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.25 μm