Merck
  • DNA double-strand break repair pathway regulates PD-L1 expression in cancer cells.
DNA double-strand break repair pathway regulates PD-L1 expression in cancer cells.
Nature communications (2017-11-25)
Hiro Sato, Atsuko Niimi, Takaaki Yasuhara, Tiara Bunga Mayang Permata, Yoshihiko Hagiwara, Mayu Isono, Endang Nuryadi, Ryota Sekine, Takahiro Oike, Sangeeta Kakoti, Yuya Yoshimoto, Kathryn D Held, Yoshiyuki Suzuki, Koji Kono, Kiyoshi Miyagawa, Takashi Nakano, Atsushi Shibata
ABSTRACT
Accumulating evidence suggests that exogenous cellular stress induces PD-L1 upregulation in cancer. A DNA double-strand break (DSB) is the most critical type of genotoxic stress, but the involvement of DSB repair in PD-L1 expression has not been investigated. Here we show that PD-L1 expression in cancer cells is upregulated in response to DSBs. This upregulation requires ATM/ATR/Chk1 kinases. Using an siRNA library targeting DSB repair genes, we discover that BRCA2 depletion enhances Chk1-dependent PD-L1 upregulation after X-rays or PARP inhibition. In addition, we show that Ku70/80 depletion substantially enhances PD-L1 upregulation after X-rays. The upregulation by Ku80 depletion requires Chk1 activation following DNA end-resection by Exonuclease 1. DSBs activate STAT1 and STAT3 signalling, and IRF1 is required for DSB-dependent PD-L1 upregulation. Thus, our findings reveal the involvement of DSB repair in PD-L1 expression and provide mechanistic insight into how PD-L1 expression is regulated after DSBs.
Sigma-Aldrich
UCN-01, ≥97% (HPLC), powder
Sigma-Aldrich
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder