Merck
  • Mutations in NEK8 link multiple organ dysplasia with altered Hippo signalling and increased c-MYC expression.
Mutations in NEK8 link multiple organ dysplasia with altered Hippo signalling and increased c-MYC expression.
Human molecular genetics (2013-02-19)
Valeska Frank, Sandra Habbig, Malte P Bartram, Tobias Eisenberger, Hermine E Veenstra-Knol, Christian Decker, Reinder A C Boorsma, Heike Göbel, Gudrun Nürnberg, Anabel Griessmann, Mareike Franke, Lori Borgal, Priyanka Kohli, Linus A Völker, Jörg Dötsch, Peter Nürnberg, Thomas Benzing, Hanno J Bolz, Colin Johnson, Erica H Gerkes, Bernhard Schermer, Carsten Bergmann
ABSTRACT
Mutations affecting the integrity and function of cilia have been identified in various genes over the last decade accounting for a group of diseases called ciliopathies. Ciliopathies display a broad spectrum of phenotypes ranging from mild manifestations to lethal combinations of multiple severe symptoms and most of them share cystic kidneys as a common feature. Our starting point was a consanguineous pedigree with three affected fetuses showing an early embryonic phenotype with enlarged cystic kidneys, liver and pancreas and developmental heart disease. By genome-wide linkage analysis, we mapped the disease locus to chromosome 17q11 and identified a homozygous nonsense mutation in NEK8/NPHP9 that encodes a kinase involved in ciliary dynamics and cell cycle progression. Missense mutations in NEK8/NPHP9 have been identified in juvenile cystic kidney jck mice and in patients suffering from nephronophthisis (NPH), an autosomal-recessive cystic kidney disease. This work confirmed a complete loss of NEK8 expression in the affected fetuses due to nonsense-mediated decay. In cultured fibroblasts derived from these fetuses, the expression of prominent polycystic kidney disease genes (PKD1 and PKD2) was decreased, whereas the oncogene c-MYC was upregulated, providing potential explanations for the observed renal phenotype. We furthermore linked NEK8 with NPHP3, another NPH protein known to cause a very similar phenotype in case of null mutations. Both proteins interact and activate the Hippo effector TAZ. Taken together, our study demonstrates that NEK8 is essential for organ development and that the complete loss of NEK8 perturbs multiple signalling pathways resulting in a severe early embryonic phenotype.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-V5 Epitope Tag Antibody, Chemicon®, from rabbit