Merck
  • Mutations in SLC39A14 disrupt manganese homeostasis and cause childhood-onset parkinsonism-dystonia.
Mutations in SLC39A14 disrupt manganese homeostasis and cause childhood-onset parkinsonism-dystonia.
Nature communications (2016-05-28)
Karin Tuschl, Esther Meyer, Leonardo E Valdivia, Ningning Zhao, Chris Dadswell, Alaa Abdul-Sada, Christina Y Hung, Michael A Simpson, W K Chong, Thomas S Jacques, Randy L Woltjer, Simon Eaton, Allison Gregory, Lynn Sanford, Eleanna Kara, Henry Houlden, Stephan M Cuno, Holger Prokisch, Lorella Valletta, Valeria Tiranti, Rasha Younis, Eamonn R Maher, John Spencer, Ania Straatman-Iwanowska, Paul Gissen, Laila A M Selim, Guillem Pintos-Morell, Wifredo Coroleu-Lletget, Shekeeb S Mohammad, Sangeetha Yoganathan, Russell C Dale, Maya Thomas, Jason Rihel, Olaf A Bodamer, Caroline A Enns, Susan J Hayflick, Peter T Clayton, Philippa B Mills, Manju A Kurian, Stephen W Wilson
ABSTRACT
Although manganese is an essential trace metal, little is known about its transport and homeostatic regulation. Here we have identified a cohort of patients with a novel autosomal recessive manganese transporter defect caused by mutations in SLC39A14. Excessive accumulation of manganese in these patients results in rapidly progressive childhood-onset parkinsonism-dystonia with distinctive brain magnetic resonance imaging appearances and neurodegenerative features on post-mortem examination. We show that mutations in SLC39A14 impair manganese transport in vitro and lead to manganese dyshomeostasis and altered locomotor activity in zebrafish with CRISPR-induced slc39a14 null mutations. Chelation with disodium calcium edetate lowers blood manganese levels in patients and can lead to striking clinical improvement. Our results demonstrate that SLC39A14 functions as a pivotal manganese transporter in vertebrates.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, HRP conjugate, Upstate®, from goat
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution