Merck
  • CCC- and WASH-mediated endosomal sorting of LDLR is required for normal clearance of circulating LDL.
CCC- and WASH-mediated endosomal sorting of LDLR is required for normal clearance of circulating LDL.
Nature communications (2016-03-12)
Paulina Bartuzi, Daniel D Billadeau, Robert Favier, Shunxing Rong, Daphne Dekker, Alina Fedoseienko, Hille Fieten, Melinde Wijers, Johannes H Levels, Nicolette Huijkman, Niels Kloosterhuis, Henk van der Molen, Gemma Brufau, Albert K Groen, Alison M Elliott, Jan Albert Kuivenhoven, Barbara Plecko, Gernot Grangl, Julie McGaughran, Jay D Horton, Ezra Burstein, Marten H Hofker, Bart van de Sluis
ABSTRACT
The low-density lipoprotein receptor (LDLR) plays a pivotal role in clearing atherogenic circulating low-density lipoprotein (LDL) cholesterol. Here we show that the COMMD/CCDC22/CCDC93 (CCC) and the Wiskott-Aldrich syndrome protein and SCAR homologue (WASH) complexes are both crucial for endosomal sorting of LDLR and for its function. We find that patients with X-linked intellectual disability caused by mutations in CCDC22 are hypercholesterolaemic, and that COMMD1-deficient dogs and liver-specific Commd1 knockout mice have elevated plasma LDL cholesterol levels. Furthermore, Commd1 depletion results in mislocalization of LDLR, accompanied by decreased LDL uptake. Increased total plasma cholesterol levels are also seen in hepatic COMMD9-deficient mice. Inactivation of the CCC-associated WASH complex causes LDLR mislocalization, increased lysosomal degradation of LDLR and impaired LDL uptake. Furthermore, a mutation in the WASH component KIAA0196 (strumpellin) is associated with hypercholesterolaemia in humans. Altogether, this study provides valuable insights into the mechanisms regulating cholesterol homeostasis and LDLR trafficking.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-HA−Peroxidase antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid