Merck
  • Autoantibody Signatures Combined with Epstein-Barr Virus Capsid Antigen-IgA as a Biomarker Panel for the Detection of Nasopharyngeal Carcinoma.
Autoantibody Signatures Combined with Epstein-Barr Virus Capsid Antigen-IgA as a Biomarker Panel for the Detection of Nasopharyngeal Carcinoma.
Cancer prevention research (Philadelphia, Pa.) (2015-05-21)
Yu-Hui Peng, Yi-Wei Xu, Li-Sheng Huang, Tian-Tian Zhai, Li-Hua Dai, Si-Qi Qiu, Yu-Su Yang, Wei-Zheng Chen, Li-Qun Zhang, En-Min Li, Li-Yan Xu
ABSTRACT
Nasopharyngeal carcinoma (NPC) is prevalent in Southern China and Southeast Asia, and autoantibody signatures may improve early detection of NPC. In this study, serum levels of autoantibodies against a panel of six tumor-associated antigens (p53, NY-ESO-1, MMP-7, Hsp70, Prx VI, and Bmi-1) and Epstein-Barr virus capsid antigen-IgA (VCA-IgA) were tested by enzyme-linked immunosorbent assay in a training set (220 NPC patients and 150 controls) and validated in a validation set (90 NPC patients and 68 controls). We used receiver-operating characteristics (ROC) to calculate diagnostic accuracy. ROC curves showed that use of these 6 autoantibody assays provided an area under curve (AUC) of 0.855 [95% confidence interval (CI), 0.818-0.892], 68.2% sensitivity, and 90.0% specificity in the training set and an AUC of 0.873 (95% CI, 0.821-0.925), 62.2% sensitivity, and 91.2% specificity in the validation set. Moreover, the autoantibody panel maintained diagnostic accuracy for VCA-IgA-negative NPC patients [0.854 (0.809-0.899), 67.8%, and 90.0% in the training set; 0.879 (0.815-0.942), 67.4%, and 91.2% in the validation set]. Importantly, combination of the autoantibody panel and VCA-IgA improved diagnostic accuracy for NPC versus controls compared with the autoantibody panel alone [0.911 (0.881-0.940), 81.4%, and 90.0% in the training set; 0.919 (0.878-0.959), 78.9%, and 91.2% in the validation set), as well as for early-stage NPC (0.944 (0.894-0.994), 87.9%, and 94.0% in the training set; 0.922 (0.808-1.000), 80.0%, and 92.6% in the validation set]. These results reveal autoantibody signatures in an optimized panel that could improve the identification of VCA-IgA-negative NPC patients, may aid screening and diagnosis of NPC, especially when combined with VCA-IgA.
Sigma-Aldrich
Nitrilotriacetic acid trisodium salt, Sigma Grade, ≥98%
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%
SAFC
Isopropyl β-D-1-thiogalactopyranoside
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening