Merck
  • Suppressor of Cytokine Signaling 3 Expression Induced by Varicella-Zoster Virus Infection Results in the Modulation of Virus Replication.
Suppressor of Cytokine Signaling 3 Expression Induced by Varicella-Zoster Virus Infection Results in the Modulation of Virus Replication.
Scandinavian journal of immunology (2015-06-16)
E-J Choi, C-H Lee, O S Shin
ABSTRACT
Varicella-zoster virus (VZV) is an important viral pathogen that is responsible for causing varicella (chickenpox) and herpes zoster (shingles). VZV has been shown to suppress early anti-viral innate immune responses, but the exact mechanisms are not yet well understood. Here we demonstrate that host control of VZV is impaired by the expression of suppressor of cytokine signaling (SOCS)3. We used three different cell types to characterize VZV-induced anti-viral and inflammatory responses. Infection of human fibroblasts (MRC-5) and human macrophages (THP-1) with VZV triggered upregulation of anti-viral responsive gene expression (IFN-α, IFN-β) in the early phases of infection, followed by the waning of these IFNs in the late phases of infection. Conversely, VZV infection in keratinocytes (HaCaT) resulted in a persistent increase in type I IFN gene expression. Interestingly, increase in SOCS1 and 3 expressions coincided with a reduction in phosphorylation of the signal transducer and activator of transcription protein 3 (STAT3) in VZV-infected MRC-5 cells. Furthermore, VZV infection increased the production of pro-inflammatory cytokines, including interleukin (IL)-6, -8, and IFN-γ-inducible protein 10 (IP-10). Knockdown of SOCS3 inhibited viral replication and enhanced secretion levels of IL-6, whereas overexpression of SOCS3 did not affect viral replication efficiency and host response. In conclusion, our data suggest that VZV infection induces SOCS3 expression, resulting in modulation of type I IFN signaling and viral replication.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human SOCS3
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Socs3