Merck
  • DNA methylation transcriptionally regulates the putative tumor cell growth suppressor ZNF677 in non-small cell lung cancers.
DNA methylation transcriptionally regulates the putative tumor cell growth suppressor ZNF677 in non-small cell lung cancers.
Oncotarget (2014-12-17)
Gerwin Heller, Corinna Altenberger, Bianca Schmid, Maximilian Marhold, Erwin Tomasich, Barbara Ziegler, Leonhard Müllauer, Christoph Minichsdorfer, György Lang, Adelheid End-Pfützenreuter, Balazs Döme, Britt-Madeleine Arns, Kwun M Fong, Casey M Wright, Ian A Yang, Walter Klepetko, Christoph C Zielinski, Sabine Zöchbauer-Müller
ABSTRACT
In our study, we investigated the role of ZNF677 in non-small cell lung cancers (NSCLC). By comparing ZNF677 expression in primary tumor (TU) and in the majority of cases also of corresponding non-malignant lung tissue (NL) samples from > 1,000 NSCLC patients, we found tumor-specific downregulation of ZNF677 expression (adjusted p-values < 0.001). We identified methylation as main mechanism for ZNF677 downregulation in NSCLC cells and we observed tumor-specific ZNF677 methylation in NSCLC patients (p < 0.0001). In the majority of TUs, ZNF677 methylation was associated with loss of ZNF677 expression. Moreover, ZNF677 overexpression in NSCLC cells was associated with reduced cell proliferation and cell migration. ZNF677 was identified to regulate expression of many genes mainly involved in growth hormone regulation and interferon signalling. Finally, patients with ZNF677 methylated TUs had a shorter overall survival compared to patients with ZNF677 not methylated TUs (p = 0.013). Overall, our results demonstrate that ZNF677 is trancriptionally regulated by methylation in NSCLCs, suggest that ZNF677 has tumor cell growth suppressing properties in NSCLCs and that ZNF677 methylation might serve as prognostic parameter in these patients.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 10% in H2O
Sigma-Aldrich
tert-Butyl acetoacetate, reagent grade, 98%
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Anti-ZNF677 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis