Merck
  • Optimization of a solid phase extraction procedure for prostaglandin E2, F2 alpha and their tissue metabolites.
Optimization of a solid phase extraction procedure for prostaglandin E2, F2 alpha and their tissue metabolites.
Prostaglandins & other lipid mediators (2007-05-15)
Toni N Welsh, Sonya Hubbard, Carolyn M Mitchell, Sam Mesiano, Pawel K Zarzycki, Tamas Zakar
ABSTRACT
The primary prostaglandins PGE(2) and PGF(2 alpha) are metabolized in tissues by a series of enzymatic and non-enzymatic reactions. To measure metabolic rates and individual reaction rates it is necessary to extract the parent prostaglandins and metabolites before the separation and quantification of each compound is achieved. Here we have established and optimized a solid phase extraction (SPE) procedure to recover PGE(2), PGF(2 alpha) and their six enzymatic and non-enzymatic tissue metabolites from aqueous solutions including urine, plasma and tissue homogenate. We have used octadecyl-bonded silica gel as the stationary phase and methanol-water mixtures as binary mobile phases. The volumes and concentrations of the washing and elution solutions were optimized individually for each PG. Recoveries of all PG standards were quantitative except for PGEM, which was recovered at 80% efficiency. Biological matrix components interfered with the extraction in a PG- and matrix-specific fashion. Inclusion of 1% formic acid in the loading mixture raised recoveries from urine, plasma and tissue homogenate to >or=90%. This SPE method is the first that has been optimized by systematic elution studies for PGE(2), PGF(2 alpha) and the complement of their tissue metabolites. The procedure is simple, robust and can serve as an effective pre-purification step before downstream separation and quantification of each tissue metabolite of PGE(2) and PGF(2 alpha) from complex biological matrices.
Product Description
Supelco
Supelclean LC-18 SPE Tube, bed wt. 500 mg, volume 6 mL, pkg of 30 ea
Supelco
Supelclean LC-18 SPE Tube, bed wt. 1 g, volume 6 mL, pk of 30