- Pindolol augmentation of antidepressant therapy.
Pindolol augmentation of antidepressant therapy.
Antidepressant therapy is not always effective and is slow to take effect. In theory, these shortfalls may be caused by induction of neuronal negative feedback via pre-synaptic 5-HTIA receptors. Pindolol, an antagonist at somatodentritic pre-synaptic 5-HTIA receptors has been investigated as a potential accelerator and augmentor of antidepressant response. A Medline search was conducted in November 1997. Six open-label studies and six controlled studies were identified for review. Open-label studies strongly suggest that pindolol may accelerate and augment antidepressant response, but controlled studies do not wholly support these findings: only three of six studies clearly demonstrate benefit. Larger, well-designed, controlled trials are needed to determine definitively the effectiveness of pindolol in this context.