Merck
Physico-chemical aspects of lactose for inhalation.
Advanced drug delivery reviews (2011-11-30)
Xiang Kou, Lai Wah Chan, Hartwig Steckel, Paul W S Heng
ABSTRACT
A dry powder inhaler (DPI) is a dosage form that consists of a powder formulation in a device which is designed to deliver an active ingredient to the respiratory tract. It has been extensively investigated over the past years and several aspects relating to device and particulate delivery mechanisms have been the focal points for debate. DPI formulations may or may not contain carrier particles but whenever a carrier is included in a commercial formulation, it is almost invariably lactose monohydrate. Many physicochemical properties of the lactose carrier particles have been reported to affect the efficiency of a DPI. A number of preparation methods have been developed which have been claimed to produce lactose carriers with characteristics which lead to improved deposition. Alongside these developments, a number of characterization methods have been developed which have been reported to be useful in the measurement of key properties of the particulate ingredients. This review describes the various physicochemical characteristics of lactose, methods of manufacturing lactose particulates and their characterization.
Product Description
Supelco
Lactose, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Lactose (anhydrous), European Pharmacopoeia (EP) Reference Standard
USP
Anhydrous lactose, United States Pharmacopeia (USP) Reference Standard