Merck
  • Influence of carbon chain length on the hepatic effects of perfluorinated fatty acids. A 19F- and 31P-NMR investigation.
Influence of carbon chain length on the hepatic effects of perfluorinated fatty acids. A 19F- and 31P-NMR investigation.
Chemical research in toxicology (1996-06-01)
C M Goecke-Flora, N V Reo
ABSTRACT
Using nuclear magnetic resonance (NMR) spectroscopy, we investigated the importance of carbon chain length with regard to the hepatic effects associated with perfluoro-n-carboxylic acids. Male F-344 rats were administered a single intraperitoneal dose of either perfluoro-n-heptanoic acid (C7-PFA), perfluoro-n-nonanoic acid (C9-PFA), or perfluoro-n-undecanoic acid (C11-PFA). Data from previous studies involving perfluoro-n-octanoic acid (C8-PFA) and perfluoro-n-decanoic acid (C10-PFA) are included for comparison. Food consumption/body weight was monitored daily for all groups. C9- and C11-PFA treatment yields a prolonged hypophagic response while C7-PFA shows a more acute response. Fluorine-19 NMR spectra of urine and bile samples show no evidence of fluorometabolites and suggest that the distribution of perfluorocarbons into urine or bile is dependent upon carbon chain length. The aqueous solubility of C7-PFA appears to facilitate rapid urinary excretion, similar to that observed for C8-PFA. The relative hydrophobicity of C9- and C11-PFA appears to favor biliary enterohepatic recirculation, yielding a more protracted toxicity, similar to C10-PFA. Phosphorus-31 NMR studies of liver in vivo and liver extracts show that perfluorocarbons of > or = C9 carbons produce a significant increase in liver phosphocholine concentration. These data are discussed with regard to the impact of these chemicals on hepatic phospholipid metabolism. Hepatic peroxisomal fatty acyl CoA-oxidase activity (FAO) was measured to determine if C7-, C9-, and C11-PFA are peroxisome proliferators. Data indicate that the induction of peroxisomal enzyme activity by perfluorocarbons requires a chain length greater than seven carbons. In general, these results demonstrate the significance of carbon chain length in the hepatotoxic response and provide clues toward understanding the processes involved in the biological activities associated with exposure to these compounds.
Sigma-Aldrich
Perfluoroundecanoic acid, 95%
Sigma-Aldrich
Perfluorononanoic acid, 97%
Sigma-Aldrich
Perfluoroheptanoic acid, ≥97.0%