Merck
  • Ginsenoside Rg1 reduces toxicity of PM(2.5) on human umbilical vein endothelial cells by upregulating intracellular antioxidative state.
Ginsenoside Rg1 reduces toxicity of PM(2.5) on human umbilical vein endothelial cells by upregulating intracellular antioxidative state.
Environmental toxicology and pharmacology (2012-12-12)
Cai-ping Li, Gang Qin, Rui-zan Shi, Ming-sheng Zhang, Ji-yuan Lv
ABSTRACT
Ambient airborne particulate matter (PM) is an important environmental pollutant responsible for many human diseases. Oxidative stress is suggested to be involved in PM-induced cell injury. The present study is designed to study unsalutary effects of the organic extracts of PM with an aerodynamic diameter of less than 2.5μm (PM(2.5)) and protective effect of Ginsenoside Rg1 (Rg1) against PM(2.5) on human umbilical vein endothelial cells (HUVECs) in vitro. Cytotoxic effects of the organic extract PM(2.5) on HUVECs were measured by means of HUVEC cell viability and the generation of intracellular reactive oxygen species (ROS). Expression of heme oxygenase-1(HO-1) and Nuclear factor-erythroid 2-related factor 2 (Nrf2) and Nrf2 cytoplasm-nucleus location were assayed. The present results showed that PM(2.5) (50-800μg/ml) decreased HUVEC viability and increased intracellular generation of ROS and malondialdehyde (MDA) in a concentration dependent manner, but increased HO-1 expression without concentration dependence. Rg1 (10 and 40μg/ml) diminished PM(2.5)-induced HUVEC viability, decrease ROS and MDA generation, increased HO-1 and Nrf2 expression and promoted Nrf2 translocation to nucleus in a concentration dependent manner. These results suggested that organic extracts of PM(2.5) increase oxidative stress and decrease cell viability; Rg1 antagonize PM(2.5)-induced excess oxidative stress; HO-1 expression increase and Nrf2 translocation to nucleus may be involved in the effects of both PM(2.5) and Rg1 on HUVECs.
Product Description
Supelco
Ginsenoside Rg1, analytical standard
Ginsenoside Rg1, primary reference standard